Learn More
P2Y6 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111893
Additional Details : Weight : 0.01000kg
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)RNRTV(S )YDLSPPILS Homology: Mouse, rat - 14/15 amino acid residues identical; human - 12/15 amino acid residues identical.
P2Y6 belongs to the family of G protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is responsive to UDP, partially responsive to UTP and ADP, and not responsive to ATP. P2Y6 expression has been documented in various blood cells, bone marrow, brain, colon, heart, kidney, ovary, placenta, spleen,and thymus. ESTs have been isolated from cervix, embryo, eye, kidney, liver/spleen, placenta, and vessel libraries.Specifications
P2Y6 (extracellular) | |
Polyclonal | |
Unconjugated | |
P2ry6 | |
2010204J23Rik; MGC15335; P2ry6; P2Y purinoceptor 6; p2y3 and chick; p2y3 receptor; P2Y6; PP2891; Purinoceptor p2y6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117264, 233571, 5031 | |
-20°C | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q15077, Q63371, Q9ERK9 | |
P2ry6 | |
Peptide corresponding to amino acid residues 172 - 186 of mouse P2RY6 (2nd extracellular loop) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |