missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p300 Monoclonal Antibody (RW109), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA116623
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: HeLa nuclear extract.
EP300 encodes the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. p300 is related by sequence to CPB (CREB-binding protein [CREB: cyclic-AMP responsive element binding protein]), and like CPB can stimulate transcription through activation of CREB. This EP300 activity is specifically inhibited by the adenovirus oncoprotein E1A. EP300 has also been identified as a co-activator of HIF1A (hypoxia-inducible factor 1 alpha), and thus plays a role in the stimulation of hypoxia-induced genes such as VEGF.Specifications
p300 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.1% sodium azide | |
B2RWS6, Q09472 | |
EP300 | |
Fusion protein containing residues 1572-2371 of human p300. | |
RUO | |
170915, 2033, 328572 | |
-20°C or -80°C if preferred | |
Liquid |
ChIP Assay, Immunocytochemistry, Immunoprecipitation, Western Blot | |
RW109 | |
Unconjugated | |
EP300 | |
A430090G16; A730011L11; E1A binding protein p300; E1A-associated protein p300; E1A-binding protein, 300kD; Ep300; Histone acetyltransferase p300; Histone butyryltransferase p300; Histone crotonyltransferase p300; KAT3B; P300; p300 HAT; Protein propionyltransferase p300; RSTS2 | |
Mouse | |
200 μL | |
Primary | |
Human, Mink, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 κ |