missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p38 MAPK beta Monoclonal Antibody (P38-11A5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 338700
Code : 50
Additional Details : Weight : 0.50000kg
Description
33-8700 has been successfully used in immunofluorescence and western blot. 33-8700 specifically recognizes the beta isoform of p38, with slight reactivity to p38-gamma.
p38 MAPK beta Monoclonal antibody specifically detects p38 MAPK beta in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
p38 MAPK beta | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Q15759, Q9WUI1 | |
MAPK11 | |
Recombinant, full-length human p38 beta-2; the product of the p38-beta gene | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1, IgG2a |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
P38-11A5 | |
Unconjugated | |
MAPK11 | |
MAP kinase 11; MAP kinase p38 beta; MAPK 11; Mapk11; mitogen activated protein kinase 11; mitogen-activated protein kinase 11; Mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38-2; OTTHUMP00000196655; p38-2; P38b; p38beta; P38BETA2; Prkm11; protein kinase, mitogen activated kinase, 11, p38beta; SAPK2; SAPK2b; Stress-activated protein kinase 2b; stress-activated protein kinase-2; stress-activated protein kinase-2b | |
Mouse | |
Protein A | |
RUO | |
19094, 5600 | |
-20°C | |
Liquid |