missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p39 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517177
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
The protein encoded by this gene is a neuron-specific activator of CDK5 kinase. It associates with CDK5 to form an active kinase. This protein and neuron-specific CDK5 activator CDK5R1/p39NCK5A both share limited similarity to cyclins, and thus may define a distinct family of cyclin-dependent kinase activating proteins.Specifications
p39 | |
Polyclonal | |
Unconjugated | |
CDK5R2 | |
B230310J22Rik; CDK5 activator 2; Cdk5r2; cyclin dependent kinase 5 regulatory subunit 2; cyclin-dependent kinase 5 activator 2; cyclin-dependent kinase 5 activator isoform p39i; cyclin-dependent kinase 5 regulatory subunit 2; cyclin-dependent kinase 5, regulatory subunit 2 (p39); NCK5AI; neuronal CDK5 activator isoform; p39; p39I; p39nck5ai; RGD1562530 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12570, 501164, 8941 | |
-20°C | |
Liquid |
Western Blot | |
20 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
O35926, Q13319 | |
CDK5R2 | |
Synthetic peptide surrounding Thr90 of human p39 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |