Learn More
P4HA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA555704
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LDYLSYAVFQ LGDLHRALEL TRRLLSLDPS HERAGGNLRY FEQLLEEERE KTLTNQTEAE LATPEGIYER PVDYLPERDV YESLCRGEGV KLTPRRQKRL FCRYHHGNRA PQLLIAPFKE EDEWDSPHIV RYYDVMS Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
This gene encodes a component of prolyl 4-hydroxylase, a key enzyme in collagen synthesis composed of two identical alpha subunits and two beta subunits. The encoded protein is one of several different types of alpha subunits and provides the major part of the catalytic site of the active enzyme. In collagen and related proteins, prolyl 4-hydroxylase catalyzes the formation of 4-hydroxyproline that is essential to the proper three-dimensional folding of newly synthesized procollagen chains. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
P4HA2 | |
Polyclonal | |
Unconjugated | |
P4HA2 | |
4-PH alpha 2; 4-PH alpha-2; 4PHalpha2; AA407196; C76437; collagen prolyl 4-hydroxylase alpha(II); C-P4Halpha(II); hypothetical protein LOC507327; P4HA2; P4hl; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha II polypeptide; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide 2; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), alpha polypeptide II; procollagen-proline,2-oxoglutarate-4-dioxygenase subunit alpha-2; prolyl 4-hydroxylase A; Prolyl 4-hydroxylase alpha IIa subunit; prolyl 4-hydroxylase subunit alpha 2; prolyl 4-hydroxylase subunit alpha-2; prolyl 4-hydroxylase, alpha polypeptide II; UNQ290/PRO330; zgc:91948 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8974 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.09 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15460 | |
P4HA2 | |
Recombinant protein corresponding to Human P4HA2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only