missing translation for 'onlineSavingsMsg'
Learn More

P5CS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen P5CS
Concentration 0.2 mg/mL
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16837664
missing translation for 'viewDocuments'
Proteintech
17719-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16827664
missing translation for 'viewDocuments'
Proteintech
17719-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene is a member of the aldehyde dehydrogenase family and encodes a bifunctional ATP- and NADPH-dependent mitochondrial enzyme with both gamma-glutamyl kinase and gamma-glutamyl phosphate reductase activities. The encoded protein catalyzes the reduction of glutamate to delta1-pyrroline-5-carboxylate, a critical step in the de novo biosynthesis of proline, ornithine and arginine. Mutations in this gene lead to hyperammonemia, hypoornithinemia, hypocitrullinemia, hypoargininemia and hypoprolinemia and may be associated with neurodegeneration, cataracts and connective tissue diseases. Alternatively spliced transcript variants, encoding different isoforms, have been described for this gene.
Specifications

Specifications

P5CS
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Human, Rat, Mouse
P54886, Q9Z110
361755, 56454, 5832
P5CS Fusion Protein Ag12100
Primary
-20°C
ALDH18A1
0.2 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ALDH18A1, GK, GPR, GSAS, P5CS, PYCS
Aldh18a1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates