missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAC 1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Procaspase-activating compound; activates procaspase-3 to produce caspase-3 (EC50 = 0.22 μM). Also activates procaspase-7 in a less efficient manner (EC50 = 4.5 μM). Pro-apoptotic; induces apoptosis in both cancerous and non-cancerous cells dependent on procaspase-3 concentration (IC50 values are 0.003 - 1.41 and 5.02 - 9.98 μM respectively).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 392.5 |
Percent Purity | >98% |
Chemical Name or Material | PAC 1 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'