Learn More
PAC 1000-7000 BumpTest Station
Brand: Draeger Safety 8318586
Code : NEW
Additional Details : Weight : 3.96000kg Transport : UN number : 1956 Chem class : 2.2
Specifications
58 L | |
Bump Test Station | |
One Test Gas Cylinder |
Safety and Handling
- divesorb pro
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P410-Protect from sunlight.
- P411-Store at temperatures not exceeding °C/°F.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide, calsium hydroxide