missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Pacific Blue™ Annexin V/SYTOX™ AADvanced™ Apoptosis Kit, for flow cytometry

Pacific Blue™ Annexin V/SYTOX™ AADvanced™ Apoptosis Kit, for flow cytometry

Brand:  Molecular Probes™ A35136

380.95 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 10118184

  • £414.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This product detects the externalization of phosphatidylserine in apoptotic cells using recombinant annexin V conjugated to violet-fluorescent Pacific Blue™ dye and dead cells using SYTOX™ AADvanced™ stain. After staining a cell population with Pacific Blue™, annexin V and SYTOX™ AADvanced™, apoptotic cells show violet fluorescence, dead cells show red fluorescence, and live cells show little or no fluorescence. These populations are easily distinguished by a flow cytometer with the 405 nm and 488 nm lines for excitation. There is very little spectral overlap between the two dyes, therefore very little compensation is needed. Each kit contains sufficient reagents for approximately 50 flow cytometry tests.

View a selection guide for all apoptosis assays for flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 kit
Apoptosis Kit
405, 488
50
Flow Cytometry
Flow Cytometer
Tube
Contains 1 vial of annexin V, Pacific Blue™ conjugate (250 μL), 1 vial of SYTOX™ AADvanced™ dead cell stain, and 1 bottle of annexin binding buffer (5X solution, 15 mL), and 1 vial of DMSO (100 μL).

Store in refrigerator (2–8°C) and protect from light.
Pacific Blue™: 415⁄455, SYTOX™ AADvanced™: 546⁄647
Wet Ice
Pacific Blue, SYTOX AADvanced Dead Cell Stain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.