missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Packaging Bags with Closure
Designed with zipper closure for storing, collecting, protecting and shipping. Buerkle™ Packaging Bags with Closure are suited for specimens, samples and sales products.
Brand: Buerkle™ 2348-0001
6.97 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Bags with a patented zipper closure. Contents remain dust-free. The film is waterproof and has good tear-resistance.- LDPE, crystal-clear
- 0.05mm strong film
- Suitable for use with foodstuff
Specifications
100 mL | |
For Storing, Collecting, Protecting and Shipping of Specimens, Samples and Sales Products and Foodstuffs | |
0.050 mm | |
130 x 60 mm | |
60 mm |
Clear | |
LDPE | |
100/Pk. | |
130 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction