Learn More
Buerkle™ Packaging Buckets
Manufactured from polypropylene for strength and durability. Buerkle™ Packaging Buckets are ideal for transport, packaging and storing of a wide range of materials.
£7.70 - £15.15
Specifications
Product Type Packaging Bucket
Material Polypropylene
Color White
For Use With (Equipment) Suitable for The Food or Building Material Industries and Non-Aggressive Chemicals in Environmental Protection, Hospitals and The Household
Includes Quality Closure and Strong Lid
View More Specs
Products 5
Product Code Brand Capacity (Metric) Diameter (Metric)Top Height (Metric) With Closure Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric)Top Height (Metric) With Closure Price Quantity & Availability  
11831091
View Documents Promotion Details
Buerkle™
2327-0001
1 L 132 mm 131 mm
£7.95
Each
In Stock
to see stock.
Edge
Add to basket
11841091
View Documents Promotion Details
Buerkle™
2327-0020
2 L 175 mm 132 mm
£7.70
Each
In Stock
to see stock.
Edge
Add to basket
11851091
View Documents Promotion Details
Buerkle™
2327-0030
3 L 225 mm 123 mm
£8.70
Each
In Stock
to see stock.
Edge
Add to basket
11861091
View Documents Promotion Details
Buerkle™
2327-0050
5 L 225 mm 194 mm
£15.15
Each
In Stock
to see stock.
Edge
Add to basket
11871101
View Documents Promotion Details
Buerkle™
2327-0010
10 L 268 mm 263 mm
£12.90
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
Suitable for food or building material industries and non-aggressive chemicals in environmental protection, hosptials and household. Features strong handle and quality closure for product safety.
  • White polypropylene
  • Air-tight closure
  • Includes lid
Specifications
Specifications
Packaging Bucket
White
Quality Closure and Strong Lid
Polypropylene
Suitable for The Food or Building Material Industries and Non-Aggressive Chemicals in Environmental Protection, Hospitals and The Household
1 Ea.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Buerkle™ Packaging Buckets