Learn More
Cytiva Packing Equipment
Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures
Brand:  Cytiva 18-1153-25
Code : GX
Additional Details : Weight : 1.00000kg
Product Code. 10808910
  • £364.00 / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
Tricorn packing equipment includes Packing equipment 5/50 (5mm connector and 50mm glass tube) or 10/100 (10mm connector and 100mm glass tube), EPDM O-rings and bottom unit, cap, and stop plug.
  • Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures. They are suitable for use with high performance media, such as Sepharose High Performance, Superdex Prep Grade, and SOURCE.
  • The columns are available with inner diameters of 5 and 10mm, and in lengths from 20 to 200mm. Column lengths of 300 and 600mm are also available for the 10mm diameter columns. The design and dimensions of the columns make them particularly useful for screening and polishing steps in protein purification.
  • Glass tubes coated with plastic film protects individuals in the laboratory from harm if the column is dropped or operated outside its specification.
Specifications
Specifications
Packing Equipment
1 x Bottom unit (cap, filter holder and O-ring), 1 x Packing tube 10/100, 1 x Packing connector
High Performance Media, Such as Sepharose High Performance, Superdex Prep Grade, and Source
1 Pack
PP/PEEK/EPDM/ Borosilicate glass
0 to 14 pH
4°C to 60°C