missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Paddle Blender Bag

Fisherbrand Paddle Blender Bags for use with Paddle Blenders.

Brand:  Fisherbrand™ IPD4400-B400

Product Code. 17359912

  • £120.00 / Pack of 500
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sterile, 400 mL bags
  • High quality, virgin LDPE
  • Resistant to rips and tears
  • Fit most paddle blenders
  • Available with or without filter
Specifications

Specifications

400 mL
Food and other samples
Paddle Blenders
300 x 180 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.