missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PageRuler™ Plus Prestained Protein Ladder, 10 to 250 kDa

Thermo Scientific™ PageRuler Plus Prestained Protein Ladder is a mixture of nine (9) blue-, orange- and green-stained proteins (10 to 250 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.

Brand:  Thermo Scientific™ 26620

406.12 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11852124

  • £572.00 / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific PageRuler Plus Prestained Protein Ladder is a mixture of nine blue-, orange-, and green-stained proteins (10 to 250 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Applications

  • Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
  • Monitoring protein transfer onto membranes after western blotting
  • Sizing of proteins on SDS-polyacrylamide gels and western blots
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contents: 10 vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Storage: Upon receipt store at -20°C

9
Yes
3 colors: Blue, Orange, Green
250, 130, 100, 70, 55, 35, 25, 15, 10 kDa
10 x 250 μL
Western Blotting, SDS-PAGE
Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm)
Protein Ladder
10 to 250 kDa
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
PageRuler™
Approved for shipment on Wet or Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.