Learn More
Thermo Scientific™ PageRuler™ Unstained Protein Ladder
A mixture of seven native proteins (14.4 to 116kDa) for use as size standards in protein electrophoresis (SDS-PAGE).
£84.00
Specifications
Number of Markers | 14 |
---|---|
Product Type | Protein Ladder |
Ready to Load | Yes |
Size Range | 10 to 200 kDa |
Stain Type | Unstained |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11802124
|
Thermo Scientific™
26614 |
2 x 250 mL |
£84.00
Pack of 2 |
In Stock
Log in to see stock. |
|||||
Description
Thermo Scientific PageRuler Unstained Protein Ladder is a mixture of 14 recombinant, highly purified, unstained proteins (10 to 200 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.
Features of the PageRuler Unstained Protein Ladder include:
- Reference bands—50 kDa band is stronger than the others for easy orientation
- Tagged—the ladder proteins contains an integral Strep-tag™ II sequence and can be detected on western blots using Strep-Tactin™ conjugates or an antibody against Strep-tag™ II Sequence
Applications
- Accurate protein sizing on SDS-polyacrylamide gels and western blots
Specifications
14 | |
Yes | |
Unstained | |
PageRuler™ |
Protein Ladder | |
10 to 200 kDa | |
200, 150, 120, 100, 85, 70, 60, 50, 40, 30, 25, 20, 15, 10 kDa |
For Research Use Only. Not for use in diagnostic procedures.