missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAIP1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | PAIP1 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897203
|
Proteintech
10675-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887203
|
Proteintech
10675-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene interacts with poly(A)-binding protein and with the cap-binding complex eIF4A. It is involved in translational initiation and protein biosynthesis. Overexpression of this gene in COS7 cells stimulates translation. Alternative splicing occurs at this locus and three transcript variants encoding three distinct isoforms have been identified.Specifications
PAIP1 | |
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Non-human Primate, Mouse, Human | |
Q8VE62, Q9H074 | |
10605, 218693, 365684 | |
PAIP1 Fusion Protein Ag1070 | |
Primary | |
-20°C | |
PAIP1 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PABP interacting protein 1, PAIP 1, PAIP1 | |
Paip1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |