missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PALB2/CEN16q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
£1235.00 - £2030.00
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Format | Tube |
For Use With (Application) | FISH-Ce |
Gene ID (Entrez) | 79728 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16053285
|
Abnova™
FG0182.100UL |
100 μL |
£1235.00
100µL |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063285
|
Abnova™
FG0182.200UL |
200 μL |
£2030.00
200µL |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark. | |
Tube | |
79728 | |
Liquid |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |