Learn More
Thermo Scientific™ PALCAM Medium
Detect and isolate Listeria monocytogenes with this differetional and selective diagnostic medium.
Brand: Thermo Scientific™ PO5104A
18.30 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Detect and isolate Listeria monocytogenes from food samples with ready-to-use Thermo Scientific™ Oxoid™ PALCAM Medium. The PALCAM Medium base is highly selective and contains a double indicator system that allows easy differentiation of Listeria monocytogenes, based upon aesculin fermentation, lack of mannitol fermentation and resultant pH change. The presence of egg yolk aids the recovery of damaged cells for presumptive identification of L. monocytogenes1.
Use PALCAM Medium for selective isolation and easy differentiation of L. monocytogenes.
- Selective: The addition of lithium chloride, ceftazidime, polymyxin B and acriflavine hydrochloride make the medium highly selective for Listeria monocytogenes.
- Easy to read: L. monocytogenes forms brown/black colonies with a black halo while mannitol positive accompanying flora, such as staphylococci and enterococci, grow as yellow colonies.
- Ready-to-use convenience of prepared media.
PALCAM Medium allows easy differential diagnosis of L. monocytogenes by utilising the double indicator system.
- Aesculin and ferrous iron
- Mannitol and phenol red
L.monocytogenes hydrolyses aesculin resulting in the formation of a black halo around colonies. L. monocytogenes does not ferment mannitol so can be easily differentiated from contaminants, such as enterococci and staphylococci, which ferment mannitolproducing a change from red to yellow in the pH indicator phenol red.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Straw | |
90mm Monoplate | |
Agar | |
10/Pk. |
2°C to 10°C | |
Germany | |
19.5g ± 1.0g |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6249171 | Certificate of Analysis | 24/03/2025 | PO5104A |
6245982 | Certificate of Analysis | 19/03/2025 | PO5104A |
6242474 | Certificate of Analysis | 10/03/2025 | PO5104A |
6239796 | Certificate of Analysis | 06/03/2025 | PO5104A |
6234234 | Certificate of Analysis | 18/02/2025 | PO5104A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.