missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium, 0.5% on 3.18mm (0.125in) alumina pellets, unreduced, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Palladium, 0.5% on 3.18mm (0.125in) alumina pellets is a heterogeneous catalyst used in chemical processes for reactions ranging from gas phase oxidation through selective hydrogenation of chemical, petrochemical, and pharmaceutical feedstocks to fuel cell power generation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 500 g |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Palladium, 0.5% on alumina pellets, 3.18mm (0.125 in.), Unreduced |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'