missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium, 10% on activated carbon paste, A402028-10, Thermo Scientific™
Click to view available options
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 7440-05-3 |
Color | Black |
Quantity | 5 g |
Molecular Formula | Pd |
MDL Number | MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 |
Synonym | black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon |
InChI Key | KDLHZDBZIXYQEI-UHFFFAOYSA-N |
SMILES | [Pd] |
IUPAC Name | palladium |
Molecular Weight (g/mol) | 106.42 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'