missing translation for 'onlineSavingsMsg'
Learn More

Palladium, 5% on activated carbon paste, 5R39, Thermo Scientific Chemicals

Debenzylation, C-N and C-O cleavage, alkene hydrogenation

£75.58 - £1422.08

Chemical Identifiers

CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11498460
View Documents
Thermo Scientific Alfa Aesar
044337.06
5 g
£75.58
5g
Please to purchase this item. Need a web account? Register with us today!
11488460
View Documents
Thermo Scientific Alfa Aesar
044337.14
25 g
£376.88
25g
Please to purchase this item. Need a web account? Register with us today!
11478460
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044337.22
100 g
£1422.08
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Palladium is used to debenzylation, C-N and C-O cleavage, alkene hydrogenation. And it is used as a catalyst for hydrogenation of alkenes, ketones, nitriles, azides, nitro groups. Used to hydrogenolysis of cyclopropanes, benzyl derivatives, halids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]

Specifications

7440-05-3
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Odorless
Black
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
Insoluble in water.
[Pd]
106.42
CHEBI:33363
Palladium, 5% on activated carbon paste, 5R39
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-115-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only