missing translation for 'onlineSavingsMsg'
Learn More

Palladium, 5% on barium sulfate powder, reduced, Thermo Scientific Chemicals

£38.09 - £1422.08

Chemical Identifiers

CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbonShow MoreShow Less
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15475985
SDS View Documents
Thermo Scientific Alfa Aesar
011722.88
2.5 g
£38.09
2.50g
Please to purchase this item. Need a web account? Register with us today!
11371808
SDS View Documents
Thermo Scientific Alfa Aesar
011722.14
25 g
£376.88
25g
Please to purchase this item. Need a web account? Register with us today!
11361808
SDS View Documents
Thermo Scientific Alfa Aesar
011722.22
100 g
£1422.08
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Palladium, 5% on barium sulfate powder reduced usually supported on Barium sulfate, or an appropriate form of carbon, and useful catalyst for hydrogenations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Palladium, 5% on barium sulfate powder, reduced, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.