missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium, 8%, Platinum, 2%; on activated carbon powder, nominally 50% water wet, Thermo Scientific™
Click to view available options
Quantity:
10 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Selective hydrogenation of nitrates to hydroxylamines, reduction of nitrogen oxides to N2 with H2 in the presence of CO and CO2
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 10 g |
Chemical Name or Material | Palladium, Platinum, Pd, 8%, Pt, 2% on activated carbon powdernominally 50% water wet |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'