missing translation for 'onlineSavingsMsg'
Learn More

Palladium on activated carbon, 10% Pd, unreduced, dry

Product Code. 10471811 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
Packaging:
Glass Bottle
Unit Size:
10g
1g
250g
50g
This item is not returnable. View return policy

Product Code. 10471811

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-05-3 | Pd | 106.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-05-3, 7440-44-0
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Color Black
Quantity 10 g
Packaging Glass Bottle
Particle Size Particle size distribution: ≤4 Micron: 10%, ≤21 Micron: 50%, ≤90 Micron: 90%
Form Powder
UN Number 3178
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Merck Index 15, 7090
Formula Weight 106.42
Percent Purity 10% (Pd)
Chemical Name or Material Palladium on Activated Carbon, 10% Pd
Show More Show Less
compliance-icons
Product Identifier
  • Palladium on activated carbon, unreduced
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P233-Keep container tightly closed.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.