missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium on activated carbon, unreduced, 5% Pd, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Quantity | 100 g |
Fieser | 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356 |
Merck Index | 15, 7090 |
Formula Weight | 106.42 |
Percent Purity | 4.8 to 5.2% (Pd, on dry basis) |
Packaging | Glass Bottle |
Particle Size | Typicaly 10% (<4 Micron) , 50% (<21 Micron), 90% (<90 Micron) |
Form | Crystals or Crystalline Powder and/or Chunks |
Chemical Name or Material | Palladium on activated carbon, 5% Pd |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
Product Identifier
- Palladium on activated carbon, unreduced
- Warning
- Flammable solid Category 2
- H228-Flammable solid.
- P233-Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'