missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium black, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used as a catalyst in hydrogenation reactions. Further it finds use in the manufacture of paints, adhesives, fibers and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Gray to Silver |
Melting Point | 1555°C |
Appearance | Powder |
Density | 12.02g/cm3 at 20°C |
Assay Percent Range | 99.9% (Metals basis) |
Quantity | 5 g |
Solubility Information | Insoluble in water; Soluble in aqua regia and fused alkalies |
Odor | Odorless |
Form | Powder |
Chemical Name or Material | Palladium black |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'