missing translation for 'onlineSavingsMsg'
Learn More

Palladium hydroxide, 99%, Thermo Scientific Chemicals

£67.13 - £1092.44

Chemical Identifiers

CAS 12135-22-7
Molecular Formula H2O2Pd
Molecular Weight (g/mol) 140.43
MDL Number MFCD00064599
InChI Key NXJCBFBQEVOTOW-UHFFFAOYSA-L
Synonym palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
PubChem CID 9942167
SMILES [OH-].[OH-].[Pd++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338298
View Documents
Thermo Scientific Alfa Aesar
039496.01
0.25 g
£67.13
0.25g
Please to purchase this item. Need a web account? Register with us today!
11348298
View Documents
Thermo Scientific Alfa Aesar
039496.03
1 g
£226.13
1g
Please to purchase this item. Need a web account? Register with us today!
11358298
View Documents
Thermo Scientific Alfa Aesar
039496.06
5 g
£1092.44
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Acts as a catalyst for arylation reactions. It is employed in hydrogenolysis of benzyl-nitrogen bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12135-22-7
140.43
NXJCBFBQEVOTOW-UHFFFAOYSA-L
9942167
H2O2Pd
MFCD00064599
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
[OH-].[OH-].[Pd++]

Specifications

12135-22-7
Powder
H2O2Pd
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
NXJCBFBQEVOTOW-UHFFFAOYSA-L
palladium(2+);dihydroxide
9942167
99%
Palladium hydroxide
Black
99%
MFCD00064599
Insoluble in water.
[OH-].[OH-].[Pd++]
140.43
140.42
Odorless
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-219-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only