missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium hydroxide on carbon, powder, unreduced, 20% Pd, moisture 50%, Thermo Scientific Chemicals
£605.01 - £2619.03
Chemical Identifiers
CAS | 12135-22-7 |
---|---|
Molecular Formula | H2O2Pd |
Molecular Weight (g/mol) | 140.43 |
MDL Number | MFCD00064599 |
InChI Key | NXJCBFBQEVOTOW-UHFFFAOYSA-L |
Synonym | palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon |
PubChem CID | 9942167 |
SMILES | [OH-].[OH-].[Pd++] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10743201
|
Thermo Scientific Acros
199620100 |
10 g | Glass bottle |
£605.01
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10002620
|
Thermo Scientific Acros
199620500 |
50 g | Glass bottle |
£2619.03
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12135-22-7 | |
140.43 | |
NXJCBFBQEVOTOW-UHFFFAOYSA-L | |
9942167 |
H2O2Pd | |
MFCD00064599 | |
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon | |
[OH-].[OH-].[Pd++] |
Specifications
12135-22-7 | |
Powder | |
Pd(OH)2 | |
16,269 | |
palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon | |
NXJCBFBQEVOTOW-UHFFFAOYSA-L | |
palladium(2+);dihydroxide | |
9942167 | |
19 to 21% (Pd, on dry basis) |
Black | |
H2O2Pd | |
MFCD00064599 | |
15, 7166 | |
Solubility in water: insoluble | |
[OH-].[OH-].[Pd++] | |
140.43 | |
140.43 | |
Palladium hydroxide on carbon, 20% Pd, moisture ca 60% |
Safety and Handling
missing translation for 'einecsNumber' : 235-219-2
missing translation for 'tsca' : TSCA
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only