Learn More
Palladium(II) 2,4-pentanedionate, Pd 34.7%, Thermo Scientific Chemicals
Description
It is used in carbonylation, oligomerization, and as a catalyst precursor for various synthetic organic transformations, for example in the preparation of complexes to efficiently catalyze Heck reaction. Useful in the decarboxylative cross-coupling of arylcarboxylic acids with aryl halides, in the preparation of monodisperse nanoparticles of CuPd alloy and in several cross-coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 210°C (decomposition) |
Color | Yellow to Orange |
Physical Form | Powder |
Quantity | 1 g |
Solubility Information | Soluble in benzene and chloroform. |
Formula Weight | 304.92 |
Concentration or Composition (by Analyte or Components) | Pd 34.7% |
Odor | Odorless |
Chemical Name or Material | Palladium(II) 2,4-pentanedionate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M15L026 | Certificate of Analysis | 17/01/2025 | 010517.03,010517.06 |
U15K029 | Certificate of Analysis | 28/08/2024 | 010517.03,010517.06 |
N19K021 | Certificate of Analysis | 05/03/2024 | 010517.03,010517.06 |
R08J042 | Certificate of Analysis | 20/02/2024 | 010517.03,010517.06 |
M09J053 | Certificate of Analysis | 17/01/2023 | 010517.03,010520.03,010517.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.