Learn More
Palladium(II) acetate, trimer, 99.98% (metals basis), Pd 47% min, Thermo Scientific Chemicals
Description
Palladium(II) acetate is a catalyst of choice for a wide variety of reactions such as vinylation, Wacker process, Buchwald-Hartwig amination, carbonylation, oxidation, rearrangement of dienes (e.g., Cope rearrangement), C-C bond formation, reductive amination, etc. Precursor to Pd(0), other Pd(II) compounds of catalytic significance, and Pd nanowires.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | ∼205°C (decomposition) |
Color | Red to Brown |
Physical Form | Powder |
Quantity | 1 g |
Linear Formula | [Pd(C2H3O2)2]3 |
Merck Index | 14,6991 |
Solubility Information | Soluble as monomer in glacial acetic acid or as trimer in benzene. |
IUPAC Name | palladium(2+) diacetate |
Formula Weight | 673.46 |
Percent Purity | 99.98% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P18L056 | Certificate of Analysis | 20/03/2025 | 043086.06,043086.03 |
Z27K063 | Certificate of Analysis | 31/12/2024 | 043086.06,043086.03 |
P22K059 | Certificate of Analysis | 31/03/2024 | 043086.06,043086.03 |
Z04J002 | Certificate of Analysis | 15/12/2023 | 043086.06,043086.03 |
N24G036 | Certificate of Analysis | 05/12/2023 | 043086.06,043086.03 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.