Learn More
Palladium(II) acetylacetonate, 35% Pd, Thermo Scientific Chemicals
£102.51 - £514.56
Chemical Identifiers
CAS | 14024-61-4 |
---|---|
Molecular Formula | C10H14O4Pd |
Molecular Weight (g/mol) | 304.64 |
MDL Number | MFCD00000025 MFCD00000025 |
InChI Key | JKDRQYIYVJVOPF-FDGPNNRMSA-L |
Synonym | palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii |
PubChem CID | 53384484 |
IUPAC Name | (E)-4-oxopent-2-en-2-olate;palladium(2+) |
SMILES | [Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10042570
|
Thermo Scientific Acros
197580010 |
1 g | Glass bottle |
£102.51
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11344112
|
Thermo Scientific Acros
197580050 |
5 g | Glass bottle |
£514.56
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14024-61-4 | |
304.64 | |
JKDRQYIYVJVOPF-FDGPNNRMSA-L | |
53384484 | |
[Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
C10H14O4Pd | |
MFCD00000025 MFCD00000025 | |
palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii | |
(E)-4-oxopent-2-en-2-olate;palladium(2+) |
Specifications
190.0°C | |
Brown-Red | |
C10H14O4Pd | |
MFCD00000025 MFCD00000025 | |
Solubility in water: insoluble in water. Other solubilities: soluble in benzene | |
[Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
304.64 | |
304.61 | |
Authentic |
14024-61-4 | |
Solution | |
(CH3COCH=C(CH3)O)2Pd | |
palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii | |
JKDRQYIYVJVOPF-FDGPNNRMSA-L | |
(E)-4-oxopent-2-en-2-olate;palladium(2+) | |
53384484 | |
35% (Pd) | |
Palladium(II) acetylacetonate, 35% Pd |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 237-859-8
missing translation for 'tsca' : TSCA
- Palladium(II) acetylacetonate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P370+P378-In case of fire: Use to extinguish.
- P420-Store away from other materials.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.