missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) bromide, Premion™, 99.99% (metals basis), Pd 39.5% min, Thermo Scientific Chemicals

Product Code. 11381958
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Palladium(II) bromide is used as an oxidizing agent, as Pd(II) catalyst and Pd(0) precatalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13444-94-5
Molecular Formula H2O2Pd
Molecular Weight (g/mol) 140.43
MDL Number MFCD00064599
InChI Key NXJCBFBQEVOTOW-UHFFFAOYSA-L
Synonym palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
PubChem CID 9942167
IUPAC Name palladium(2+) dihydroxide
SMILES [OH-].[OH-].[Pd++]

Specifications

Melting Point (decomposition)
Color Black
Physical Form Powder
Quantity 2 g
Assay Percent Range 99.99% (metal basis)
Solubility Information soluble in hydrogen bromide. Insoluble in water.
Formula Weight 266.22
Odor Odorless
Sensitivity Hygroscopic
Appearance Black moist powder
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Chemical Name or Material Palladium hydroxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'