missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) chloride, 99.9% (metals basis), Pd 59.0% min, Thermo Scientific Chemicals

For Use As a Catalyst Precursor

POA - £1687.40

Chemical Identifiers

CAS 7647-10-1
Molecular Formula Cl2Pd
Molecular Weight (g/mol) 177.32
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
PubChem CID 24290
ChEBI CHEBI:53434
SMILES [Cl-].[Cl-].[Pd++]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11391658
View Documents
Thermo Scientific Alfa Aesar
011034.02
0.5 g
£90.65
0.50g
Please to purchase this item. Need a web account? Register with us today!
11381658
View Documents
Thermo Scientific Alfa Aesar
011034.04
2 g
£362.81
2g
Please to purchase this item. Need a web account? Register with us today!
11361658
View Documents
Thermo Scientific Alfa Aesar
011034.09
10 g
£1687.40
10g
Please to purchase this item. Need a web account? Register with us today!
11371658
View Documents
Thermo Scientific Alfa Aesar
011034.18
50 g
POA
50g
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

Catalyst precursor. Palladium(II) chloride is used to remove the stain present in stainless steel . Its solutions are used to test the corrosion-resistance of stainless steel. It is used in carbon monoxide detectors. It is also used for cosmetic tattooing of leukomas in the cornea and in the preparation of palladium(II) iodide by reacting with potassium iodide. It is involved in the synthesis of semiconducting metal-containing polymers. It is used as a catalyst for carbonylation of organic tellurides and deamination of phenethylamines to phenyl substituted pyrroles with copper(I) as co catalyst. It plays an important role in the hydrodimerization of butadiene to form octa-2,7-dien-1-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-10-1
177.32
PIBWKRNGBLPSSY-UHFFFAOYSA-L
CHEBI:53434
Cl2Pd
MFCD00003558
24290
[Cl-].[Cl-].[Pd++]

Specifications

675°C (decomposition)
Red to Brown
99.9% (metals basis)
MFCD00003558
Soluble in dilute mineral acids,aqueous metal halides.Soluble in cold water,organic solvents and hydrochloric acid.
[Cl-].[Cl-].[Pd++]
177.32
CHEBI:53434
Odorless
4 g/cm3 at 20°C
7647-10-1
Crystalline
Cl2Pd
palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
PIBWKRNGBLPSSY-UHFFFAOYSA-L
dichloropalladium
24290
177.31
Red-brown powder
Palladium(II) chloride
Videos
Safety and Handling

Safety and Handling

P234-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P390-P501c

H290-H302-H317-H318

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'einecsNumber' : 231-596-2

missing translation for 'rtecsNumber' : RT3500000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.