Learn More
Palladium(II) nitrate hydrate, 99.8% (metals basis), Pd 39% min, Thermo Scientific Chemicals
Description
Palladium(II) nitrate hydrate is used as a precursor to supported palladium catalysts. It is also used as a catalyst in the conversion of alkene nitration to glycol dinitrates. It is involved in the separation of chlorine and iodine and used as biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | (decomposition) |
Color | Brown to Red |
Physical Form | Crystalline |
Quantity | 2 g |
Linear Formula | Pd(NO3)2·xH2O |
UN Number | UN1477 |
Merck Index | 14,6992 |
Solubility Information | Soluble in dilute nitric acid. |
Formula Weight | 230.43 (Anhydrous) |
Percent Purity | 99.8% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N13L023 | Certificate of Analysis | 17/02/2025 | 011035.09,011035.18,011035.04 |
N05L016 | Certificate of Analysis | 07/02/2025 | 011035.09,011035.18,011035.04 |
Y15K014 | Certificate of Analysis | 19/11/2024 | 011035.09,011035.18,011035.04 |
Y01K036 | Certificate of Analysis | 14/11/2024 | 011035.09,011035.18,011035.04 |
U21K003 | Certificate of Analysis | 17/09/2024 | 011035.09,011035.18,011035.04 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.