Learn More
Palladium(II) nitrate, solution, Pd 4-5% w/w (cont. Pd), Thermo Scientific Chemicals
Description
Palladium(II) nitrate dehydrate is used as a reactant for doping activated carbon for catalysis. It acts as a catalyst for hydrogenation of 1-heptyne and naphthalene. It serves as an important raw material for the preparation of platinum-palladium carbon alloy nanocatalysts for methanol-tolerant oxygen reduction reaction in fuel cells and in the synthesis of copper palladium alloy thin films. Furthermore, it is used in the synthesis of di-phenyl sulfide-modified palladium-titanium dioxide catalysts for acetylene hydrogenation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Brown to Orange |
Linear Formula | Pd(NO3)2 |
UN Number | UN3264 |
Concentration | Pd 4 to 5% w/w (cont. Pd) |
Formula Weight | 230.43 |
Quantity | 1 g |
Physical Form | Liquid |
Chemical Name or Material | Palladium (II) nitrate solution |
RUO – Research Use Only