missing translation for 'onlineSavingsMsg'
Learn More
Palladium(II) oxide, anhydrous, 99.9% (metals basis), Pd 85% min, Thermo Scientific Chemicals Product Code.: 11311678

Palladium(II) oxide, anhydrous, 99.9% (metals basis), Pd 85% min, Thermo Scientific Chemicals

Product Code. 11311678
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11311678

Brand: Thermo Scientific Alfa Aesar 011040.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Catalytic reduction, in organic synthesis

Catalytic reduction, in organic synthesis.Palladium(II) oxide is used as a catalyst for reduction in organic synthesis. It is an excellent gas sensing material for carbon monoxide at low working temperatures. It is also used in the preparation of good optical quality films. It is involved in the preparation of LuPd2O4 , which is used in superconducting materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-08-5
Molecular Formula OPd
Molecular Weight (g/mol) 122.42
MDL Number MFCD00011172
InChI Key JQPTYAILLJKUCY-UHFFFAOYSA-N
Synonym palladium ii oxide, palladium oxide pdo, acmc-1bqvh, palladium oxide, pdo, palladium ii oxide, anhydrous, palladium ii oxide-pd, 11113-77-2 cpd with unspecified mf, palladium oxide ? inverted exclamation mark-?, palladium ii oxide trace metals basisShow MoreShow Less
PubChem CID 5083724
SMILES [O--].[Pd++]

Specifications

Melting Point 870°C
Color Brown
Physical Form Powder
Quantity 5 g
Merck Index 14,6993
Solubility Information Slightly soluble in aqua regia; Insoluble in water and acids.
IUPAC Name palladium(2+) oxidandiide
Formula Weight 122.4
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pd 85% min.
Odor Odorless
Density 8.70 g/mL
Chemical Name or Material Palladium(II) oxide, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX24K047 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number011040.06,011040.03
Lot NumberP25K002 Certificate TypeCertificate of Analysis Date05/04/2024 Catalog Number011040.06,011040.03
Lot NumberZ09I010 Certificate TypeCertificate of Analysis Date09/01/2023 Catalog Number011040.06,011041.09,011040.03
Lot NumberF16N18 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011040.06,011041.09,011040.03
Lot NumberM25C058 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011040.06,011041.09,011040.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Palladium(II) oxide, anhydrous, 99.9% (metals basis), Pd 85% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.