missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium Silver foil, 0.5mm (0.02in) thick, 99.9% (metals basis excluding Pt), Thermo Scientific Chemicals
£787.00 - £2705.00
Specifications
Chemical Name or Material | Palladium Silver foil |
---|---|
Name Note | 0.5mm (0.02 in.) thick |
Avg. Mol. Wt. or Mol. Wt. Range | Pd:Ag; 75:25 wt% |
Assay Percent Notes | (metals basis excluding Pt) |
MDL Number | MFCD02091738 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11325427
|
Thermo Scientific Alfa Aesar
042684.FF |
25 x 25 mm |
£787.00
25mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346139
|
Thermo Scientific Alfa Aesar
042684.FI |
50 x 50 mm |
£2705.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Palladium silver foils are used in dentistry for dental crowns and bridges. It also used in conductive films and pastes. It is used to make multilayer capacitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Palladium Silver foil | |
Pd:Ag; 75:25 wt% | |
MFCD02091738 | |
99.9% | |
≈3.64g/25x25mm |
0.5mm (0.02 in.) thick | |
(metals basis excluding Pt) | |
Insoluble in water. | |
Odorless | |
Foil |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only