missing translation for 'onlineSavingsMsg'
Learn More
Palladium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 15444775

Palladium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15444775
5 Pc.
Click to view available options
Quantity:
1 Pc.
5 Pc.
This item is not returnable. View return policy

Product Code. 15444775

Brand: Thermo Scientific Alfa Aesar 040418.KJ

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

For Use As Evaporation Source, Hydrogen Getter, Jewelry Production

Evaporation source, hydrogen getter, jewelry production, electrical contacts for semiconductorsPalladium slug finds application in jewelry production and in electrical contacts for semiconductors. It is also used as a hydrogen getter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Form: Approx.2.41g/slug; Material made from precision cut of palladium wire
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Appearance Silver gray rod
Density 12.02g/cm3 at 20°C
Assay Percent Range 99.9% (Metals basis)
Quantity 5 Pc.
Percent Purity 99.9%
Form Slug
Assay (metals basis)
Chemical Name or Material Palladium, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Palladium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.