Learn More
Palladium standard solution, for AAS, 1 mg/ml Pd in 10% HCl, Thermo Scientific Chemicals
£40.40 - £147.74
Specifications
Chemical Name or Material | Palladium standard solution |
---|---|
Concentration | 997 to 1003 μg/mL of Pd (2+) in 10% HCl (20°C) |
Name Note | 1 mg/ml Pd in 10% HCl |
CAS | 7440-05-3, 7647-01-0, 7732-18-5 |
Packaging | Plastic bottle |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10472581
|
Thermo Scientific Acros
196171000 |
100 mL | Plastic bottle |
£40.40
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10685642
|
Thermo Scientific Acros
196175000 |
500 mL | Plastic bottle |
£147.74
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Palladium standard solution | |
1 mg/ml Pd in 10% HCl | |
Plastic bottle | |
Pd | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
23938 | |
AAS | |
-25.0°C | |
1.05 | |
Solution |
997 to 1003 μg/mL of Pd (2+) in 10% HCl (20°C) | |
7440-05-3, 7647-01-0, 7732-18-5 | |
Brown/Red | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
[Pd] | |
106.42 | |
CHEBI:33363 | |
1.0500g/mL | |
107.0°C | |
Soluble in water |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May be corrosive to metals.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep only in original container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- Palladium atomic absorption standard solution
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.