missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palmatine chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Quantity | 250 mg |
Formula Weight | 387.86 |
MDL Number | MFCD00016656 |
Physical Form | Crystalline Powder |
Color | Light-yellow to Yellow to Orange |
Purity | ≥97.5% |
InChI Key | RLQYRXCUPVKSAW-UHFFFAOYSA-M |
SMILES | [Cl-].COC1=CC=C2C=C3C4=CC(OC)=C(OC)C=C4CC[N+]3=CC2=C1OC |
IUPAC Name | 3,4,10,11-tetramethoxy-7,8-dihydro-6λ⁵-azatetraphen-6-ylium chloride |
Molecular Weight (g/mol) | 387.86 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'