missing translation for 'onlineSavingsMsg'
Learn More

Palmitic acid hydrazide, 98+%, Thermo Scientific Chemicals

Product Code. 11397836
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2619-88-7
Molecular Formula C16H34N2O
Molecular Weight (g/mol) 270.461
MDL Number MFCD00066357
InChI Key SSVSELJXJJCANX-UHFFFAOYSA-N
Synonym palmitohydrazide, palmitic acid hydrazide, hexadecanoyl hydrazide, hexadecanoic acid hydrazide, palmiticacidhydrazide, hexadecanohydrazide #, acmc-1cbma, 16-hexadecanoyl hydrazide, hexadecanoic acid,hydrazide, hexdecanoic acid hydrazide
PubChem CID 225536
IUPAC Name hexadecanehydrazide
SMILES CCCCCCCCCCCCCCCC(=O)NN

Specifications

Melting Point 150°C to 152°C
Linear Formula CH3(CH2)14CONHNH2
Quantity 5 g
Beilstein 1784616
Formula Weight 270.46
Percent Purity 99%
Chemical Name or Material Palmitic acid hydrazide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'