missing translation for 'onlineSavingsMsg'
Learn More

Palmitoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11494884
Click to view available options
Quantity:
100 g
500 g

Palmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Palmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.

Solubility
Soluble in ether.Decomposes in water and alcohol.

Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents, Bases, Alcohols.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 112-67-4
Molecular Formula C16H31ClO
Molecular Weight (g/mol) 274.873
MDL Number MFCD00000742
InChI Key ARBOVOVUTSQWSS-UHFFFAOYSA-N
Synonym palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride
PubChem CID 8206
IUPAC Name hexadecanoyl chloride
SMILES CCCCCCCCCCCCCCCC(=O)Cl

Specifica

Melting Point ∼9°C to 12°C
Density 0.906
Boiling Point 174°C (0.5 mmHg)
Flash Point 182°C (359°F)
Linear Formula CH3(CH2)14COCl
Refractive Index 1.452
Quantity 100 g
UN Number UN3265
Beilstein 972409
Sensitivity Moisture sensitive
Solubility Information Soluble in ether.Decomposes in water and alcohol.
Formula Weight 274.88
Percent Purity 98%
Chemical Name or Material Palmitoyl chloride
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'