Learn More
Palmitoyl chloride, 98%, Thermo Scientific Chemicals
£54.40 - £193.00
Chemical Identifiers
CAS | 112-67-4 |
---|---|
Molecular Formula | C16H31ClO |
Molecular Weight (g/mol) | 274.873 |
MDL Number | MFCD00000742 |
InChI Key | ARBOVOVUTSQWSS-UHFFFAOYSA-N |
Synonym | palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride |
PubChem CID | 8206 |
IUPAC Name | hexadecanoyl chloride |
SMILES | CCCCCCCCCCCCCCCC(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494884
|
Thermo Scientific Alfa Aesar
A13812.22 |
100 g |
£54.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404894
|
Thermo Scientific Alfa Aesar
A13812.36 |
500 g |
£193.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Palmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPalmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.
Solubility
Soluble in ether.Decomposes in water and alcohol.
Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents, Bases, Alcohols.
Chemical Identifiers
112-67-4 | |
274.873 | |
ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
8206 | |
CCCCCCCCCCCCCCCC(=O)Cl |
C16H31ClO | |
MFCD00000742 | |
palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
hexadecanoyl chloride |
Specifications
112-67-4 | |
0.906 | |
182°C (359°F) | |
CH3(CH2)14COCl | |
MFCD00000742 | |
972409 | |
palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride | |
ARBOVOVUTSQWSS-UHFFFAOYSA-N | |
hexadecanoyl chloride | |
8206 | |
98% |
∼9°C to 12°C | |
174°C (0.5 mmHg) | |
C16H31ClO | |
1.452 | |
UN3265 | |
Moisture sensitive | |
Soluble in ether.Decomposes in water and alcohol. | |
CCCCCCCCCCCCCCCC(=O)Cl | |
274.873 | |
274.88 | |
Palmitoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H314-H317
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 203-996-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only