missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palmityl palmitate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used as emollients, masking agents and skin conditioning and a glosser and thickener for creams. It improves emulsion texture and stability and gives structure to cosmetic sticks. Used in skin and hair applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as emollients, masking agents and skin conditioning and a glosser and thickener for creams. It improves emulsion texture and stability and gives structure to cosmetic sticks. Used in skin and hair applications.
Solubility
Soluble in hot acetone.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
It is used as emollients, masking agents and skin conditioning and a glosser and thickener for creams. It improves emulsion texture and stability and gives structure to cosmetic sticks. Used in skin and hair applications.
Solubility
Soluble in hot acetone.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 55°C to 56°C |
Linear Formula | CH3(CH2)14CO2(CH2)15CH |
Quantity | 1 g |
Solubility Information | Soluble in hot acetone. |
Formula Weight | 480.85 |
Percent Purity | 96% |
Chemical Name or Material | Palmityl palmitate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'