Learn More
Description
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.
Solubility
Soluble in nitrobenzene, pyridine
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | >300°C |
Assay Percent Range | 99% |
Beilstein | 901319 |
Merck Index | 14,7005 |
Solubility Information | Soluble in nitrobenzene, pyridine |
Formula Weight | 388.38 |
Percent Purity | 99% |
Quantity | 50 g |
Chemical Name or Material | Pamoic Acid |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0462880 | Certificate of Analysis | 30/08/2024 | A15207.30,A15207.18,A15207.0B |
A0440874 | Certificate of Analysis | 22/04/2024 | A15207.30,A15207.18,A15207.0B |
GBFA030284 | Certificate of Analysis | 22/11/2021 | A15207.30,A15207.18,A15207.0B |
F5811A | Certificate of Analysis | 22/11/2021 | A15207.30,A15207.18,A15207.0B |
FA030284 | Certificate of Analysis | 22/11/2021 | A15207.30,A15207.18,A15207.0B |
Safety and Handling

- Pamoic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Pamoic acid
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.