missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Panoramasque™ Full-Face Air Purifying Respirator (APR)
The Panoramasque™ Full-Face APR is a Mono filter with Rd40 thread connection system that offers respiratory protection, depending on the canister type assembled on it.
Brand: Honeywell™ 1711000
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Product Type: Full-Face Air Purifying Respirator (APR)
- Material (Body): EPDM or Silicone
- Material (Lens): PMMA, Glass, Chlorine Hazard Specific
- Color: Black, Yellow
- Cartridge: Single Filter via Rd40 (1/7 in.) Connection
- Use with: EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters
- PPE Class: Variable (Dependant on Cartridge)
- Hazard Types: Particulates, Gas, Vapor, Smoke
- Applications: Chemical Industry, Oil and Gas Industries, Pharmaceutical Industry
Specifications
EN 136:1989 | |
Black | |
Full-Face Air Purifying Respirator (APR) | |
Chemical Industry, Oil and Gas Industries, Pharmaceutical Industry | |
Five-Point Harness | |
Specific for Chlorine Hazards |
1/7 in. Rd40 Thread System | |
Mono-Filter | |
EPDM | |
EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters | |
Without Speech Diaphragm | |
Yes |