missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Panoramasque™ Full-Face Air Purifying Respirator (APR)

The Panoramasque™ Full-Face APR is a Mono filter with Rd40 thread connection system that offers respiratory protection, depending on the canister type assembled on it.

Brand:  Honeywell™ 1711001

 View more versions of this product

Product Code. 12672923

  • £383.00 / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Product Type: Full-Face Air Purifying Respirator (APR)
  • Material (Body): EPDM or Silicone
  • Material (Lens): PMMA, Glass, Chlorine Hazard Specific
  • Color: Black, Yellow
  • Cartridge: Single Filter via Rd40 (1/7 in.) Connection
  • Use with: EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters
  • PPE Class: Variable (Dependant on Cartridge)
  • Hazard Types: Particulates, Gas, Vapor, Smoke
  • Applications: Chemical Industry, Oil and Gas Industries, Pharmaceutical Industry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EN 136:1989
Black
Full-Face Air Purifying Respirator (APR)
Chemical Industry, Oil and Gas Industries, Pharmaceutical Industry
Five-Point Harness
Laminated Glass for Chemical Hazards
1/7 in. Rd40 Thread System
Mono-Filter
EPDM
EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters
Without Speech Diaphragm
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers