missing translation for 'onlineSavingsMsg'
Learn More
Pantoprazole sodium salt hydrate, Thermo Scientific Chemicals Product Code.: 15557539

Pantoprazole sodium salt hydrate, Thermo Scientific Chemicals

Product Code. 15557539
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15557539

Brand: Thermo Scientific Acros 460870050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 718635-09-7
Molecular Formula C16H14F2N3NaO4S
Molecular Weight (g/mol) 405.35
MDL Number MFCD08704580
InChI Key YNWDKZIIWCEDEE-UHFFFAOYNA-N
Synonym pantoprazole sodium, pantoprazole sodium hydrate, pantozol hydrate, protonix hydrate, c16h14f2n3nao4s.h2o, sodium pantoprazole 1-hydrate, pantoprazole sodium hydrate hplc, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methanesulfinyl-1-sodio-1,3-benzodiazole hydrate, 5-difluoromethoxy-2-3,4-dimethoxy-2-pyridinyl methyl sulfinyl-1h-benzimidazole sodium salt hydrate
PubChem CID 23684923
IUPAC Name sodium;5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methylsulfinyl]benzimidazol-1-ide;hydrate
SMILES [Na+].COC1=C(OC)C(CS(=O)C2=NC3=CC(OC(F)F)=CC=C3[N-]2)=NC=C1

Specifications

Melting Point 151.0°C to 155.0°C
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information (1% in water) Clear solution
Formula Weight 405.35
Chemical Name or Material Pantoprazole sodium salt hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458160 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number460870010,460870050
Lot NumberA0455726 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number460870010,460870050
Lot NumberA0453037 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number460870010,460870050
Lot NumberA0403426 Certificate TypeCertificate of Analysis Date21/06/2021 Catalog Number460870010,460870050
compliance-icons
Product Identifier
  • Pantoprazole sodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pantoprazole sodium salt hydrate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.