missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Papain, for Biochemistry

Product Code. 10400832 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10g
100g
500g
Unit Size:
100g
10g
500g
This item is not returnable. View return policy

Product Code. 10400832

Brand: Thermo Scientific Chemicals 416761000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Name Papain
pH Range 4.8 to 6.2
Quantity 100g
Source Plant
Packaging Glass bottle
Form Powder
What does the unit TU/mg mean for enzyme activity versus the commonly used unit U/mg?

Papain activity is measured in terms of units of tyrosine release per mg (TU/mg), and this unit can be used interchangeably with the more general units per mg (U/mg).

compliance-icons
Product Identifier
  • Papain
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.