Learn More
Epredia™ Papanicolaou EA Stains
Control the color hues and intensities of your work with Epredia™ Papanicolaou Stain EAs, useful for both gynecological and non-gynecological staining.
Brand:  Epredia™ 75611
Code : LE
Additional Details : Weight : 3.79000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
 View more versions of this product
Product Code. 10540529
  • £77.55 / Each
Estimated Shipment: 01-10-2024
to see stock.
Description
Description
  • EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
  • EA-65 is used with OG-6 for non-gynecological staining.
  • Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.
Specifications
Specifications
EA-65 Papanicolaou Stain
EA-65; 3.79L (1 Gal.)
1 gal.